Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®
Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Magnesium chloride hexahydrate 99.0-101.0%, crystals FCC, USP, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets F.C.C. Requirements:
    99.0 - 101.0%
  • Meets U.S.P Requirements:
    Passes Test
  • Food GMP Manufactured Product:
    Passes Test
  • GMP Manufactured Product:
    4.5 - 7.0
  • Preserve in Tight Containers:
    ≤0.005%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.005%
  • Bulk Food Chemical:
    Passes Test
  • Bulk Pharmaceutical Chemical:
    ≤0.01%
  • USP - Assay:
    99.0 - 101.0 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - pH (1 in 20):
    4.5 - 7.0
  • USP - Insoluble Matter:
    ≤0.005 %
  • USP - Sulfate (SO₄):
    ≤0.005 %
  • USP - Barium (Ba):
    Passes Test
  • USP - Calcium (Ca):
    ≤0.01 %
  • USP - Potassium (K):
    Passes Test
  • FCC - Assay:
    99.0 - 101.0 %
  • FCC - Ammonium (NH₄):
    ≤0.005 %
  • FCC - Sulfate (SO₄):
    ≤0.005 %
  • FCC - Lead (Pb):
    ≤4 mg/kg

Specification Test Results

Related Information

Frequently Bought Together