Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium chloride hexahydrate 99-102%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Magnesium chloride hexahydrate 99-102%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Magnesium chloride hexahydrate 99-102%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    99.0 - 102.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.005%
  • Assay (MgCl₂·6H₂O):
    99.0 - 102.0 %
  • Insoluble Matter:
    ≤0.005 %
  • pH of 5% Solution at 25°C:
    4.5 - 7.0
  • Nitrate (NO₃):
    ≤0.001 %
  • Sulfate (SO₄):
    ≤0.002 %
  • Ammonium (NH₄):
    ≤0.002 %
  • Barium (Ba):
    ≤0.005 %
  • Calcium (Ca):
    ≤0.01 %
  • Potassium (K):
    ≤0.005 %
  • Sodium (Na):
    ≤0.005 %
  • Strontium (Sr):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Manganese (Mn):
    ≤5.0 ppm

Specification Test Results

Related Information

Frequently Bought Together