Hydrogen peroxide 30%, Finyte® for microelectronic, J.T.Baker®
Catalog # 5816-09
Supplier: Avantor
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:500 lb
- Seq:7323
Specifications
Specification Test Results
- Assay (H₂O₂):30.0 - 32.0 %
- Color (APHA):≤ 6
- Free Acid (µeq/g):≤ 0.6
- Residue after Evaporation:≤ 5 ppm
- Chloride (Cl):≤ 0.06 ppm
- Nitrate (NO₃):≤ 0.4 ppm
- Phosphate (PO₄):≤ 0.20 ppm
- Sulfate (SO₄):≤ 0.2 ppm
- Trace Impurities - Aluminum (Al):≤ 10.0 ppb
- Arsenic and Antimony (as As):≤ 5.0 ppb
- Trace Impurities - Barium (Ba):≤ 10.0 ppb
- Trace Impurities - Beryllium (Be):≤ 10.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 10.0 ppb
- Trace Impurities - Boron (B):≤ 10.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 10.0 ppb
- Trace Impurities - Calcium (Ca):≤ 10.0 ppb
- Trace Impurities - Chromium (Cr):≤ 10.0 ppb
- Trace Impurities - Cobalt (Co):≤ 10.0 ppb
- Trace Impurities - Copper (Cu):≤ 10.0 ppb
- Trace Impurities - Gallium (Ga):≤ 10.0 ppb
- Trace Impurities - Germanium (Ge):≤ 10.0 ppb
- Trace Impurities - Gold (Au):≤ 10.0 ppb
- Trace Impurities - Iron (Fe):≤ 10.0 ppb
- Trace Impurities - Lead (Pb):≤ 10.0 ppb
- Trace Impurities - Lithium (Li):≤ 10.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 10.0 ppb
- Trace Impurities - Manganese (Mn):≤ 10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 10.0 ppb
- Trace Impurities - Nickel (Ni):≤ 10.0 ppb
- Trace Impurities - Niobium (Nb):≤ 10.0 ppb
- Trace Impurities - Potassium (K):≤ 10.0 ppb
- Trace Impurities - Silicon (Si):≤ 10.0 ppb
- Trace Impurities - Silver (Ag):≤ 10.0 ppb
- Trace Impurities - Sodium (Na):≤ 10.0 ppb
- Trace Impurities - Strontium (Sr):≤ 10.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 10.0 ppb
- Trace Impurities - Thallium (Tl):≤ 10.0 ppb
- Trace Impurities - Tin (Sn):≤ 10.0 ppb
- Trace Impurities - Titanium (Ti):≤ 10.0 ppb
- Trace Impurities - Vanadium (V):≤ 10.0 ppb
- Trace Impurities - Zinc (Zn):≤ 10.0 ppb
- Trace Impurities - Zirconium (Zr):≤ 10.0 ppb
- Particle Count - 0.2 µm and greater:≤ 1175 par/ml
- Particle Count at point of fill - 0.5 µm and greater:≤ 150 par/ml
Specification Test Results
About this item
:
ADR:5.1,II
:Management of Change (MOC) category = R
:34.01 g/mol
UN:2014
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333