Order Entry
Czech Republic
ContactUsLinkComponent
Hydrogen peroxide 30%, Finyte® for microelectronic use, J.T.Baker®
Oxidizing
Corrosive
Toxic
Health
Hydrogen peroxide 30%, Finyte® for microelectronic use, J.T.Baker®
Catalog # 5516-07
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Corrosive
Toxic
Health
Hydrogen peroxide 30%, Finyte® for microelectronic use, J.T.Baker®
Catalog # 5516-07
Supplier:  Avantor
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    HDPE
  • Pk:
    19 L
  • Seq:
    5282

Specifications

Specification Test Results

  • For Microelectronic Use:
  • Assay (H₂O₂):
    30.0 - 32.0 %
  • Color (APHA):
    ≤ 6
  • Free Acid (µeq/g):
    ≤ 0.6
  • Residue after Evaporation:
    ≤ 5 ppm
  • Chloride (Cl):
    ≤ 0.06 ppm
  • Nitrate (NO₃):
    ≤ 0.4 ppm
  • Phosphate (PO₄):
    ≤ 0.20 ppm
  • Sulfate (SO₄):
    ≤ 0.2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤ 10.0 ppb
  • Arsenic and Antimony (as As):
    ≤ 5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤ 10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤ 10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤ 10.0 ppb
  • Trace Impurities - Boron (B):
    ≤ 10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤ 10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤ 10.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤ 10.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤ 10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤ 10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤ 10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤ 10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤ 10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤ 10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤ 10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤ 10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤ 10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤ 10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤ 10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤ 10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤ 10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤ 10.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤ 10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤ 10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤ 10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤ 10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤ 10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤ 10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤ 10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤ 10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤ 10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤ 10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤ 10.0 ppb
  • Particle Count - 0.2 µm and greater:
    ≤ 1175 par/ml
  • Particle Count - 0.5 µm and greater, For Information Only:
    For Information Only

Specification Test Results

About this item

 :
ADR:5.1,II
 :Management of Change (MOC) category = R
 :34.01 g/mol
UN:2014
 :GHS03
 :GHS05
 :GHS06
 :GHS08
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333