Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Irritant
Hydrochloric acid 25%, Biotech Reagent, J.T.Baker®
Catalog # 0323-07
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid 25%, Biotech Reagent, J.T.Baker®
Catalog # 0323-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Jerrican, poly pail
  • Pk:
    19 L
  • Seq:
    5282

Specifications

Specification Test Results

  • GMP Manufactured Product:
    24.0 - 26.0%
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.0100%
  • Appearance (Clear, colorless):
    Passes Test
  • Assay (HCl):
    24.0 - 26.0 %
  • Residue after Evaporation:
    ≤ 0.0100 %
  • Arsenic (As):
    ≤ 1.500 ppm
  • Sulfate (SO₄):
    ≤ 1 ppm
  • Ammonium (NH₄):
    ≤ 3 ppm
  • Heavy Metals (as Pb):
    ≤ 0.5 ppm
  • Iron (Fe):
    ≤ 0.1 ppm

Specification Test Results

About this item

 :
ADR:8,II
 :Filtered through a 0.2 µm filter.
 :36.46 g/mol
UN:1789
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324