Cyclohexane, AR®, Laboratory Reagent, Macron Fine Chemicals™
Catalog # 5412-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:84.16 g/mol
UN:1145
ADR:3,II
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814