Order Entry
Czech Republic
ContactUsLinkComponent
Chloroform stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Chloroform stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5285.2500
Supplier:  Avantor
CAS Number:  
undefined
Chloroform stabilised, BAKER ANALYZED® for pesticide residue analysis, J.T.Baker®
Catalog # 5285.2500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (by GC):
    min. 99.8 %
  • Acidity:
    max. 0.0005 meq/g
  • Evaporation Residue:
    max. 5 ppm
  • GC sensitive impurities (lindane GC-ECD):
    max. 5 ng/l
  • Preservative (Ethanol):
    about 1%
  • Water (KF):
    max. 0.05 %

Specification Test Results

About this item

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
 :
 :119.38 g/mol
UN:1888
 :GHS08
 :GHS07
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826
Boiling Pt:61.7
 :∼ 1% Stabilised with ethanol
Melting Pt:-63
ADR:6.1 III
Merck Index:13,02160