Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Irritant
Environment
Ammonia solution 20%, BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Catalog # 9380.0500
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Environment
Ammonia solution 20%, BAKER INSTRA-ANALYZED® Plus for trace metal analysis, J.T.Baker®
Catalog # 9380.0500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • —:
  • For Laboratory,Research,or Manufacturing Use:
  • Color (APHA):
    ≤10
  • Assay (as NH₃):
    20 - 22 %
  • Chloride (Cl):
    ≤0.5 ppm
  • Phosphate (PO₄):
    ≤0.01 ppm
  • Sulfate (SO₄):
    ≤1 ppm
  • Trace Impurities - Aluminum (Al):
    ≤1 ppb
  • Trace Impurities - Antimony (Sb):
    ≤0.5 ppb
  • Trace Impurities - Arsenic (As):
    ≤1 ppb
  • Trace Impurities - Barium (Ba):
    ≤0.1 ppb
  • Trace Impurities - Beryllium (Be):
    ≤0.1 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤0.1 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤0.5 ppb
  • Trace Impurities - Calcium (Ca):
    ≤1 ppb
  • Trace Impurities - Cerium (Ce):
    ≤0.1 ppb
  • Trace Impurities - Cesium (Cs):
    ≤0.1 ppb
  • Trace Impurities - Chromium (Cr):
    ≤0.5 ppb
  • Trace Impurities - Cobalt (Co):
    ≤0.5 ppb
  • Trace Impurities - Copper (Cu):
    ≤0.5 ppb
  • Trace Impurities - Dysprosium (Dy):
    ≤0.1 ppb
  • Trace Impurities - Erbium (Er):
    ≤0.1 ppb
  • Trace Impurities - Europium (Eu):
    ≤0.1 ppb
  • Trace Impurities - Gadolinium (Gd):
    ≤0.1 ppb
  • Trace Impurities - Gallium (Ga):
    ≤0.1 ppb
  • Trace Impurities - Germanium (Ge):
    ≤0.1 ppb
  • Trace Impurities - Gold (Au):
    ≤0.5 ppb
  • Trace Impurities - Hafnium (Hf):
  • Trace Impurities - Holmium (Ho):
    ≤0.1 ppb
  • Trace Impurities - Indium (In):
    ≤0.1 ppb
  • Trace Impurities - Iron (Fe):
    ≤1 ppb
  • Trace Impurities - Lanthanum (La):
    ≤0.1 ppb
  • Trace Impurities - Lead (Pb):
    ≤0.1 ppb
  • Trace Impurities - Lithium (Li):
    ≤0.1 ppb
  • Trace Impurities - Lutetium (Lu):
    ≤0.1 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤1 ppb
  • Trace Impurities - Manganese (Mn):
    ≤0.5 ppb
  • Trace Impurities - Mercury (Hg):
    ≤0.2 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤0.5 ppb
  • Trace Impurities - Neodymium (Nd):
    ≤0.1 ppb
  • Trace Impurities - Nickel (Ni):
    ≤0.5 ppb
  • Trace Impurities - Niobium (Nb):
    ≤0.1 ppb
  • Trace Impurities - Palladium (Pd):
  • Trace Impurities - Platinum (Pt):
  • Trace Impurities - Potassium (K):
    ≤1 ppb
  • Trace Impurities - Praseodymium (Pr):
    ≤0.1 ppb
  • Trace Impurities - Rhenium (Re):
  • Trace Impurities - Rhodium (Rh):
    ≤0.5 ppb
  • Trace Impurities - Rubidium (Rb):
    ≤0.1 ppb
  • Trace Impurities - Ruthenium (Ru):
  • Trace Impurities - Samarium (Sm):
    ≤0.1 ppb
  • Trace Impurities - Scandium (Sc):
    ≤0.1 ppb
  • Trace Impurities - Selenium (Se):
    ≤1 ppb
  • Trace Impurities - Silver (Ag):
    ≤0.5 ppb
  • Trace Impurities - Sodium (Na):
    ≤1 ppb
  • Trace Impurities - Strontium (Sr):
    ≤0.1 ppb
  • Trace Impurities - Tellurium (Te):
    ≤0.1 ppb
  • Trace Impurities - Terbium (Tb):
    ≤0.1 ppb
  • Trace Impurities - Thallium (Tl):
    ≤0.1 ppb
  • Trace Impurities - Thorium (Th):
    ≤0.1 ppb
  • Trace Impurities - Thulium (Tm):
    ≤0.1 ppb
  • Trace Impurities - Tin (Sn):
    ≤0.5 ppb
  • Trace Impurities - Titanium (Ti):
    ≤0.5 ppb
  • Trace Impurities - Tungsten (W):
    ≤0.1 ppb
  • Trace Impurities - Uranium (U):
    ≤0.1 ppb
  • Trace Impurities - Vanadium (V):
    ≤0.5 ppb
  • Trace Impurities - Ytterbium (Yb):
    ≤0.1 ppb
  • Trace Impurities - Yttrium (Y):
    ≤0.1 ppb
  • Trace Impurities - Zinc (Zn):
    ≤0.5 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤0.1 ppb

Specification Test Results

About this item

 :
 :35.05 g/mol
UN:2672
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650