Order Entry
Czech Republic
ContactUsLinkComponent
Diammonium hydrogen orthophosphate, BAKER ANALYZED® ACS, J.T.Baker®
Diammonium hydrogen orthophosphate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0054.1000
Supplier:  Avantor
CAS Number:  
Diammonium hydrogen orthophosphate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0054.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 98.0 %
  • Calcium (Ca):
    max. 0.001 %
  • Iron (Fe):
    max. 0.001 %
  • Potassium (K):
    max. 0.005 %
  • Magnesium (Mg):
    max. 0.0005 %
  • Sodium (Na):
    max. 0.005 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Insoluble Matter:
    max. 0.005 %
  • Nitrate (NO3):
    max. 0.003 %
  • pH of 5% Solution at 25°C:
    7.7 - 8.1
  • Sulfate (SO4):
    max. 0.01 %
  • Trace Impurities (in ppm)Chloride (Cl):
    max. 5

Specification Test Results

About this item

 :
 :132.06 g/mol
CAS Number:7783-28-0
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891