Order Entry
Czech Republic
ContactUsLinkComponent
Ammonium carbonate ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™
Ammonium carbonate ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™
Catalog # 3352-20
CAS Number:  
undefined
Ammonium carbonate ≥30% (NH₃ basis), AR® ACS, Macron Fine Chemicals™
Catalog # 3352-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    = 30 %
  • ACS - Assay (as NH₃):
    ≥30 %
  • ACS - Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Nonvolatile Matter:
    ≤0.01 %
  • ACS - Sulfur Compounds (as SO₄):
    ≤0.002 %

Specification Test Results

About this item

 :
 :96.09 g/mol
Merck Index:13,00508
Melting Pt:58 °C
Density:1.5 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-058-0
CAS Number:10361-29-2
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890