


Kyselina octová ledová 99.7-100.5% USP, FCC, ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:64-19-7
:
Formula:H₃CCOOH
MDL Number:MFCD00036152
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1,05 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
REACH:01-2119475328-30
:GHS02
:GHS05
:GHS07
Specification Test Results
- GMP Manufactured Product:99.7 - 100.5%
- Meets ACS Reagent Chemical Requirements:Passes Test
- Meets F.C.C. Requirements:≥15.6 °C
- Food GMP Manufactured Product:≤5 ppm
- Meets U.S.P Requirements:Passes Test
- IMPORTANT: Material will freeze if stored below 17 °C (63°F).:≤0.005%
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Food Chemical:Passes Test
- Bulk Pharmaceutical Chemical:99.7 - 100.5%
- USP - Assay (CH₃COOH):99.7 - 100.5 %
- USP - Chloride (Cl) (USP):Passes Test
- USP - Congealing Temperature:≥15.6 °C
- USP - Identification:Passes Test
- USP - Nonvolatile Residue:≤0.005 %
- USP - Readily Oxidizable Substances:Passes Test
- USP - Sulfate (USP)(SO₄):Passes Test
- FCC - Assay (CH₃COOH):99.7 - 100.5 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤0.5 mg/kg
- FCC - Nonvolatile Residue:≤0.005 %
- FCC - Readily Oxidizable Substances:Passes Test
- FCC - Solidification Point:≥15.6 °C
- ACS - Assay (CH₃COOH) (by GC):≥99.7 %
- ACS - Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- ACS - Chloride (Cl):≤1 ppm
- ACS - Color (APHA):≤10
- Dilution Test:Passes Test
- ACS - Heavy Metals (as Pb) (ACS):≤0.5 ppm
- Trace Impurities - ACS - Iron (Fe):≤0.200 ppm
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Substances Reducing Dichromate (ml):≤0.40
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Sulfate (SO₄):≤1 ppm
- ACS - Titrable Base (meq/g):≤0.0004
- Arsenic (As):≤3 ppm
- Copper (Cu):≤0.00001 %
- Freezing Point:≥16.0 °C
- Nickel (Ni):≤0.00001 %
- Appearance:Passes Test
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR2502
SDS
spec_MACR2502-14 spec_MACR2502-21 spec_MACR2502-27 spec_MACR2502-46
Specifikace produktu
Frequently Bought Together