Order Entry
Czech Republic
ContactUsLinkComponent
 
EDTA disodium salt dihydrate 99.0-101.0%, AR® ACS, Macron Fine Chemicals™
Catalog # 4931-20
CAS Number:  
undefined
EDTA disodium salt dihydrate 99.0-101.0%, AR® ACS, Macron Fine Chemicals™
Catalog # 4931-20
CAS Number:  

Specifications

  • Pack type:
    Case, Mall STAKMOR
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
    99.0 - 101.0%
  • ACS - Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):
    99.0 - 101.0 %
  • Identification A:
    Passes Test
  • Identification B:
    Passes Test
  • Identification C:
    Passes Test
  • ACS - Heavy Metals (as Pb):
    ≤0.005 %
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤0.01 %
  • ACS - Nitrilotriacetic Acid (by polarography):
    ≤0.1 %
  • ACS - pH of 5% Solution at 25°C:
    4.0 - 6.0

Specification Test Results

About this item

 :
 :372.24 g/mol
Merck Index:13,03543
Melting Pt:255 °C
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037