Order Entry
Czech Republic
ContactUsLinkComponent
Kovac's indole reagent for microbiology
Corrosive
Flammable
Irritant
Kovac's indole reagent for microbiology
Catalog # APPC252908.1608
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Corrosive
Flammable
Irritant
Kovac's indole reagent for microbiology
Catalog # APPC252908.1608
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :149.19 g/mol
Storage Temperature:Refrigerator
UN:2920
ADR:8 (3),II
 :GHS05
 :GHS02
 :GHS07
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381