Order Entry
Czech Republic
ContactUsLinkComponent
1-Octanol ≥99% ACS, Riedel-de Haën™
Irritant
1-Octanol ≥99% ACS, Riedel-de Haën™
Certificates
undefined
Irritant
1-Octanol ≥99% ACS, Riedel-de Haën™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Non-volatile matter:
    Max. 0.004 %
  • Titrable Acid:
    Max. 0.0002 meq/g
  • APHA:
    Max. 10
  • Refractive Index:
    n20/D 1.429 (lit.)
  • Vapor Pressure:
    0.14 mmHg ( 25 °C)
  • Autoignition Temperature:
    523 °F

Specification Test Results

Related Information

Frequently Bought Together