Order Entry
Czech Republic
ContactUsLinkComponent
Titanium(IV) oxide sulphate sulphuric acid hydrate solution (27 - 31% H₂SO₄ basis), Millipore®
Corrosive
Titanium(IV) oxide sulphate sulphuric acid hydrate solution (27 - 31% H₂SO₄ basis), Millipore®
Catalog # SIAL89532-1L
Supplier:  Merck
CAS Number:  
Corrosive
Titanium(IV) oxide sulphate sulphuric acid hydrate solution (27 - 31% H₂SO₄ basis), Millipore®
Catalog # SIAL89532-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Density:
    1,32 g/ml at 20 °C
  • Assay:
    27 - 31% H₂SO₄ basis ~5% Ti basis

Specification Test Results

About this item

 :
 :GHS05
CAS Number:123334-00-9
 :MFCD00011270
Formula:TiOSO₄·xH₂SO₄·yH₂O
MDL Number:MFCD00150565