Reagent for volumetric KF titration, HYDRANAL™
Supplier: Honeywell Chemicals
Certificates
Certificates
About this item
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
Potassium permanganate 0.05 mol/l (0.25 N) in aqueous solution
Supplier: VWR Chemicals
Lab soakers, Versi-Dry® and Super Versi-Dry®, Nalgene®
Supplier: Thermo Fisher Scientific
Dishes, disposable
Supplier: Korff
Seals for threaded connection caps, BOLA
Supplier: Bohlender
Pipette tips, Standard
Supplier: Brand
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Benzoic acid, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Reagent for volumetric KF titration, HYDRANAL™
Supplier: Honeywell Chemicals
Disodium sulphide 90+%, pure
Supplier: Thermo Fisher Scientific
Sodium hydroxide, micropearls, extra pure
Supplier: Thermo Fisher Scientific
Avantor® ACE® Phenyl, HPLC Columns, 3 µm
Supplier: Avantor
Methyl orange 0.1% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Karl Fischer reagents, buffer substances, HYDRANAL™, Fluka™
Supplier: Honeywell Chemicals
Accessories for Retsch sieves
Supplier: Retsch GmbH
Desiccator plates
Supplier: Brand
Stoppers, rubber, conical
Supplier: KARTELL