Anti-KRT8 Mouse Monoclonal Antibody [clone: C-51]
ABNOMAB14664
: Abnova
- Antibody type:Primary
- Antigen name:Cytoskeleton preparation from HeLa cells.
- Antigen symbol:KRT8
- Clonality:Monoclonal
- Clone:C-51
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3856|3875
- Immunogen:Cytoskeleton preparation from HeLa cells.
- Purification:Protein G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against native human KRT8 and KRT18.
Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: C-51
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human