Anti-dsDNA Mouse Monoclonal Antibody [clone: 121-3]
Catalog # ABNOMAB14666
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclei of Burkitt's cells
- Antigen symbol:dsDNA
- Clonality:Monoclonal
- Clone:121-3
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Form:Liquid
- Immunogen:Nuclei of Burkitt's cells.
- Purification:Protein G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against human double stranded DNA.
Type: Primary
Antigen: dsDNA
Clonality: Monoclonal
Clone: 121-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human