Anti-CD81 Mouse Monoclonal Antibody (APC-C750) [clone: MEM-38]
Catalog # ABNOMAB15373
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD81 molecule
- Antigen symbol:CD81
- Clonality:Monoclonal
- Clone:MEM-38
- Conjugation:APC-C750
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:975
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C.
- Immunogen:MOLT-4 (human T-ALL cell line).
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human CD81.
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: MEM-38
Conjugation: APC-C750
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human