Order Entry
Czech Republic
ContactUsLinkComponent
Chicken embryo extract, MP Biomedicals
Chicken embryo extract, MP Biomedicals
Catalog # ICNA092850145
Supplier:  MP Biomedicals
undefined
Chicken embryo extract, MP Biomedicals
Catalog # ICNA092850145
Supplier:  MP Biomedicals

Specifications

  • Format:
    Liquid
  • Colour:
    Light brown
  • Description:
    Chicken embro extract
  • Storage conditions:
    −20 °C
  • Application:
    Mammalian cell culture; Neurostem cell culture
  • Pk:
    20 mL

Specifications

About this item

Chicken embryo extract prepared from nine to 12 day old eggs.

  • Soluble in water
  • Brown solution with dirty aspect
  • Collected from registered flocks, which are free from clinical signs of notifiable diseases

Chicken embryo extract is prepared from 9 to 12 day old eggs collected from registered flocks and found to be free from clinical signs of notifiable diseases.

Delivery: Supplied in Dulbecco's PBS.