Order Entry
Czech Republic
ContactUsLinkComponent
Genomic DNA purification kits, Fast DNA™ kit, MP Biomedicals
Genomic DNA purification kits, Fast DNA™ kit, MP Biomedicals
Catalog # ICNA116540400
Supplier:  MP Biomedicals
CAS Number:  
Genomic DNA purification kits, Fast DNA™ kit, MP Biomedicals
Catalog # ICNA116540400
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Yeast/Fungi
    Cultured cells
    Invertebrates
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    FastDNA™ Kit, 100×2 ml preps
  • Description:
    FastDNA™ Kit

Specifications

About this item

The FastDNA™ Kit quickly and efficiently isolates high quality genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin filter method. It can be used with the FastPrep®-24 or FastPrep® FP120 instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.

Lysing Matrix A and three different chaotropic buffers are provided for the optimal homogenisation of a wide variety of sample types. The released DNA is purified by a silica-based GeneClean® SPIN procedure, with supplied filter buckets and catch tubes to simplify liquid handling. Isolated DNA is suitable for PCR analysis and other downstream applications.

Delivery: The kit includes Lysing Matrix A (100× 2 ml tubes), ¼" ceramic spheres (100 spheres), Binding Matrix (66 ml), concentrated SEWS-M (12 ml), DES (25 ml), CLS VF (90 ml), PPS (25 ml), CLS-TC (110 ml) and CLS-Y (110 ml).