ContactUsLinkComponent
Dehydrated culture media, Terrific broth II, MP Biomedicals
Dehydrated culture media, Terrific broth II, MP Biomedicals
Katalogové číslo ICNA113046075
Supplier:  MP Biomedicals
CAS Number:  
Dehydrated culture media, Terrific broth II, MP Biomedicals
Katalogové číslo ICNA113046075
Supplier:  MP Biomedicals
CAS Number:  
Specifications
  • Pk:
    10 L
  • Description:
    Terrific broth II, pack pouches
  • Seq:
    07
  • Volume:
    10000 ml
Specifications
About this item
Terrific broth (TB) has been formulated for optimum growth of Escherichia coli for purposes of strain maintenance, cloning, protein expression. It is certified for plasmid and other bacterial cultures in liquid medium. Higher cell densities are achieved using this broth.
  • Molecular biology certified bacterial medium
Terrific Broth II is a proprietary formulation that is shipped as a single powder instead of as two separate components exhibiting same growth characteristics as the original terrific broth formula.