Order Entry
Czech Republic
ContactUsLinkComponent
Tetrahydroxy-1,4-benzoquinone hydrate ≥99%, Sigma-Aldrich®
Irritant
Tetrahydroxy-1,4-benzoquinone hydrate ≥99%, Sigma-Aldrich®
Catalog # SIALT17000-5G
Supplier:  Merck
CAS Number:  
Irritant
Tetrahydroxy-1,4-benzoquinone hydrate ≥99%, Sigma-Aldrich®
Catalog # SIALT17000-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :190.11 g/mol
Melting Pt:>300 °C (lit.)
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:123334-16-7
Formula:C₆H₆O₇
MDL Number:MFCD00001597