Goggles, GoggleGear™ 500, Scotchgard™
111-2327
: 3M
- Type:Anti-UV, anti-fog, anti-scratch
- Colour:Red/black
- Lenses:Clear PC
- Pk:1
The vented goggles Gear 500 Scotchgard™ have a low-profile design with an adjustable elastic strap, a low profile frame and a PC lens.
- Optical class 1, excellent UV protection
- Longlasting Scotchgard™ Anti-fog coating prevends lens from fogging and scratches
- Coating withstands disinfection with diluted bleach soaking or alcohol wipes without losing its anti-fog performance
For use with prescription inserts.
: EN 166:2001 ; 2C-1.2 3M 1 BT K N