Order Entry
Czech Republic
ContactUsLinkComponent
DNA purification kits, GeneClean® for Ancient DNA™ Kit, MP Biomedicals
DNA purification kits, GeneClean® for Ancient DNA™ Kit, MP Biomedicals
Catalog # ICNA111002200
Supplier:  MP Biomedicals
CAS Number:  
undefined
DNA purification kits, GeneClean® for Ancient DNA™ Kit, MP Biomedicals
Catalog # ICNA111002200
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    GeneClean® for Ancient DNA™ Kit
  • Description:
    GeneClean® for Ancient DNA™ Kit

Specifications

About this item

The GeneClean® for Ancient DNA™ Kit is ideal for purification of fragmented or damaged DNA from aged or preserved samples. It is designed for isolation of up to 20 μg DNA from bone, preserved tissue, soil organisms, or animal by-products.

GeneClean® for Ancient DNA™ Kit minimises the possibility of contamination by contemporary DNA from extraneous sources. All solutions are prepared from ultra-pure reagents and dispensed in a dedicated laminar flow hood.