Order Entry
Czech Republic
ContactUsLinkComponent
Cleanroom mops, King
Cleanroom mops, King
Catalog # PPSA2140020
undefined
Cleanroom mops, King
Catalog # PPSA2140020

Specifications

  • Description:
    King, best tuft quality, 150 g
  • Length:
    400 mm
  • Pk:
    1

Specifications

About this item

Mops with high area performance.

  • Provides maximum surface capacity and excellent dirt pick-up capability
  • Reference in the single or dual-stage process
  • Ready-to-use pre-washed
  • Made up of synthetic/viscose yarn (50:50)
  • With cover sheet, PVC pocket and sewed up double-latching pocket