Plastic hydrometers, Durac™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Made from optical quality polycarbonate, these plain form hydrometers are designed for applications where rough handling or contamination could be an issue.
- Durable and shatterproof, lead free ballast
- Easy to read black scale on yellow back to avoid eye fatigue
- Individually serialised − Temperature standardisation of 15 °C
Suitable for applications in the food or beverage industries.
Certifications: Traceable to National Institute of Standards and Technology (NIST).
Delivery: Includes a multi-language statement of accuracy.
Caution: Not recommended for use with petroleum-based liquids or other corrosives, these should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
Frequently Bought Together