Order Entry
Czech Republic
ContactUsLinkComponent
Zinc 1000 µg/ml, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Irritant
Zinc 1000 µg/ml, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Catalog # 6946.0100
Supplier:  Avantor
CAS Number:  
Irritant
Zinc 1000 µg/ml, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Catalog # 6946.0100
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :65.39 g/mol
 :GHS07
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291