Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD5 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 53-7.3]
Anti-CD5 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 53-7.3]
Catalog # ABNOMAB15309
Supplier:  Abnova
Anti-CD5 Rat Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 53-7.3]
Catalog # ABNOMAB15309
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD5 molecule
  • Antigen symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    53-7.3
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    12507
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4°C.
  • Immunogen:
    Mouse thymocytes/splenocytes.
  • Purification:
    Affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal antibody raised against mouse Cd5.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse