Order Entry
Czech Republic
ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ Activated Spherical Carbon, SPE Columns
J.T.Baker® BAKERBOND spe™ Activated Spherical Carbon, SPE Columns
Catalog # 7536-00
Supplier:  Avantor
J.T.Baker® BAKERBOND spe™ Activated Spherical Carbon, SPE Columns
Catalog # 7536-00
Supplier:  Avantor

Specifications

  • Capacity:
    6 ml
  • Seq:
    01
  • Bed weight:
    1000 mg
  • Phase:
    Activated spherical carbon
  • Pk:
    100 g

Specifications

About this item

These are disposable extraction columns packed with activated spherical carbon. Designed for the extraction and purification of organic compounds.