ContactUsLinkComponent
Ultrasonic pipette cleaners
Ultrasonic pipette cleaners
Certificates
undefined
Ultrasonic pipette cleaners
Specifications
  • Catalog No:
  • BAND2070
  • Model:
  • PR 140 DH
  • Capacity:
  • 18 L
  • Max. output power:
  • 800 W
  • W×D×H:
  • 150×150×895 mm