Order Entry
Czech Republic
ContactUsLinkComponent
 
Cleanroom mops, MicroMop
Catalog # 141-1392
undefined
Cleanroom mops, MicroMop
Catalog # 141-1392

Specifications

  • Description:
    MicroMop, 120 g
  • Length:
    400 mm
  • Pk:
    1

Specifications

About this item

Mop for all smooth hard floorings. Made up of 100% PES super microfiber.

  • For wet and damp cleaning in the single-step wiping process
  • Also suitable for dry-cleaning, sealed parquet and laminate flooring
  • Backing with cover sheet and belt bag extremely tear