

Zinc stearate 12.5 - 14.0 % RSN, Hyqual®, Macron Fine Chemicals™
Catalog # 4644-32
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Poly bag
- Pk:50 lb
- Seq:7137
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:12.5 - 14.0 %
- Assay (Zn(C₁₈H₃₅O₂)₂):12.5 - 14.0 %
- Appearance:Passes Test
- Identification A:Passes Test
- Identification B:≥54 °C
- Arsenic (As):≤1.500 ppm
- Loss on Drying:≤1.25 %
- Lead (Pb):≤0.001 %
- Alkalies and Alkaline Earths (as SO₄):≤1.0 %
- Thru U.S. No. 325 Sieve:≥99.0 %
Specification Test Results
About this item
:
:632.34 g/mol
Density:1.10 g/cm³
Flash Pt:279.00 °C
:GHS07
:GHS09
CAS Number:557-05-1
Formula:C₃₆H₇₀O₄Zn
MDL Number:MFCD00013031