Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-GPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-GPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-9133R-A350
Supplier:  Bioss
Anti-GPS1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-9133R-A350
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    G Protein Pathway Suppressor 1
  • Symbol antigenu:
    GPS1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Alexa Fluor® 350
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Křížová adsorbce:
    No
  • Formulář:
    liquid
  • Identifikační číslo genu:
    2873
  • Synonyma antigenu:
    COPS1|GPS1|CSN1|Arabidopsis FUS6/COP11 homolog|G protein pathway suppressor 1|MGC71287
  • Modifikace:
    phospho-Ser474
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Skladovací teplota:
    Store at –20 °C for 12 months
  • Koncentrace:
    1 μg/μl
  • Transportní teplota:
    4°C
  • Čištění:
    Purified by Protein A
  • Bal.:
    100 µl

Specifications

About this item

Essential component of the COP9 signalosome complex (CSN), a complex involved in various cellular and developmental processes. The CSN complex is an essential regulator of the ubiquitin (Ubl) conjugation pathway by mediating the deneddylation of the cullin subunits of SCF-type E3 ligase complexes, leading to decrease the Ubl ligase activity of SCF-type complexes such as SCF, CSA or DDB2. The complex is also involved in phosphorylation of p53/TP53, c-jun/JUN, IkappaBalpha/NFKBIA, ITPK1 and IRF8/ICSBP, possibly via its association with CK2 and PKD kinases. CSN-dependent phosphorylation of TP53 and JUN promotes and protects degradation by the Ubl system, respectively. Suppresses G-protein-and mitogen-activated protein kinase-mediated signal transduction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat