Specifikace
- Antibody type:Primary
- Antigen name:Arylsulfatase H
- Antigen symbol:ARSH
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:347527
- Antigen synonyms:Arylsulfatase H|ARSH_HUMAN.|ASH
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifikace
O tomto produktu
Sulfatases hydrolyze sulfate esters from sulfated steroids, carbohydrates, proteoglycans and glycolipids. They are involved in hormone biosynthesis, modulation of cell signaling and degradation of macromolecules. Arylsulfatase H, also known as ARSH, is a 562 amino acid protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Localized to the plasma membrane, Arylsulfatase H uses calcium as a cofactor to hydrolyze sulfate esters. The gene encoding Arylsulfatase D maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ARSH
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human