Anti-KRT1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-9080R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Keratin 1
- Antigen symbol:KRT1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3848
- Antigen synonyms:KRT1A|NEPPK|EHK|CK1|K1|EHK1|EPPK
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. It can phosphorylate a large number of proteins. Participates in Wnt signaling.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: KRT1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat