Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CARKD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-CARKD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-9402R-A350
Supplier:  Bioss
Anti-CARKD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-9402R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Carbohydrate Kinase Domain Containing
  • Antigen symbol:
    CARKD
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    Carbohydrate kinase domain containing|LP3298|NNRD_HUMAN.|carbohydrate kinase domain-containing protein|FLJ10769
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

CARKD is a 347 amino acid protein that belongs to the YjeF family and exists as three alternatively spliced isoforms. Containing one YjeF C-terminal domain, CARKD is encoded by a gene that maps to human chromosome 13q34. Chromosome 13 contains around 114 million base pairs and 400 genes. Key tumor suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. RB1 encodes a crucial tumor suppressor protein which, when defective, leads to malignant growth in the retina and has been implicated in a variety of other cancers. The gene SLITRK1, which is associated with Tourette syndrome, is on chromosome 13. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival. Trisomy 13, also known as Patau syndrome, is quite deadly and the few who survive past one year suffer from permanent neurologic defects, difficulty eating and vulnerability to serious respiratory infections.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CARKD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat