Anti-RBM20 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-9606R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RNA Binding Motif Protein 20
- Antigen symbol:RBM20
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:282996
- Antigen synonyms:RNA-binding motif protein 20|RNA-binding protein 20|RBM20
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:402-448/1227
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
RNA-binding protein that acts as a regulator of mRNA splicing of a subset of genes involved in cardiac development. Regulates splicing of TTN (Titin).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RBM20
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat