Order Entry
Czech Republic
ContactUsLinkComponent
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-9517R-A647
Supplier:  Bioss
Anti-AMPD3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-9517R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AMP deaminase 3
  • Antigen symbol:
    AMPD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    272
  • Antigen synonyms:
    AMP deaminase 3|Erythrocyte AMP deaminase|AMPD3|AMP deaminase isoform E
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

AMP deaminase plays a critical role in energy metabolism.Involvement in diseaseDefects in AMPD3 are the cause of adenosine monophosphate deaminase deficiency erythrocyte type (AMPDDE); also known as erythrocyte AMP deaminase deficiency. AMPDDE is a metabolic disorder due to lack of activity of the erythrocyte isoform of AMP deaminase. It is a clinically asymptomatic condition characterized by a 50% increase in steady-state levels of ATP in affected cells. Individuals with complete deficiency of erythrocyte AMP deaminase are healthy and have no hematologic disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMPD3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat