Anti-LHPP Rabbit Polyclonal Antibody (Cy3®)
BOSSBS-8319R-CY3
: Bioss
- Antibody type:Primary
- Antigen name:Phospholysine Phosphohistidine Inorganic Pyrophosphate Phosphatase
- Antigen symbol:LHPP
- Clonality:Polyclonal
- Conjugation:Cy3®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:64077
- Antigen synonyms:HDHD2B
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
LHPP, also known as HDHD2B, is a 270 amino acid protein that exists as a homodimer and is a member of the HAD-like hydrolase superfamily. Expressed in liver, kidney and moderately in brain, LHPP is encoded by a gene located on 10, which houses over 1,200 genes and comprises nearly 4.5% of the human genome. Defects in some of the genes that map to chromosome 10 are associated with Charcot-Marie Tooth disease, Jackson-Weiss syndrome, Usher syndrome, nonsyndromatic deafness, Wolman’s syndrome, Cowden syndrome, multiple endocrine neoplasia type 2 and porphyria.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LHPP
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat