Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GTF2H1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-GTF2H1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-8175R-A350
Supplier:  Bioss
Anti-GTF2H1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-8175R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    General Transcription Factor 2H1
  • Antigen symbol:
    GTF2H1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2965
  • Antigen synonyms:
    Basic transcription factor 2 62 kDa subunit|P62|BTF2|Basic transcription factor 62 kDa subunit|TFB1|BTF2 p62|General transcription factor IIH polypeptide 1|gtf2h1|General transcription factor IIH subunit 1|BTF 2|TFIIH|TFIIH basal transcription factor complex p62 subunit|General transcription factor IIH,polypeptide 1,62kDa|TF2H1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    230-280/548
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTF2H1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat