Anti-SIPA1L2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-7927R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Signal-induced Proliferation-associated 1 Like 2
- Antigen symbol:SIPA1L2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:57568
- Antigen synonyms:Signal induced proliferation associated 1 like protein 2|Signal-induced proliferation-associated 1-like protein 2|SI1L2_HUMAN|SIPA1 like protein 2|SIPA1-like protein 2|SIPA1L2|SPAL2|KIAA1389.
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
SIPA1L2
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SIPA1L2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat