Anti-CCNDBP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Katalogové číslo BOSSBS-7885R-A350
Dodavatel: Bioss
Specifikace
- Antibody type:Primary
- Antigen name:Cyclin D-type Binding-protein 1
- Antigen symbol:CCNDBP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:23582
- Antigen synonyms:GCIP|grap2 and cyclin-D-interacting protein|DIP1|human homolog of Maid|cyclin-D1-binding protein 1|cyclin D-type binding-protein 1|HHM
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifikace
O tomto produktu
May negatively regulate cell cycle progression. May act at least in part via inhibition of the cyclin-D1/CDK4 complex, thereby preventing phosphorylation of RB1 and blocking E2F-dependent transcription.Tissue specificity: Ubiquitously expressed. Expression is down-regulated in a variety of tumor types including breast, colon, prostate and rectal tumors, and is up-regulated in certain hepatic carcinomas.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCNDBP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat