Anti-NUBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-7818R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nucleotide Binding Protein 2
- Antigen symbol:NUBP2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10101
- Antigen synonyms:CFD1|NBP 2|NUBP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Component of the cytosolic iron-sulfur (Fe/S) protein assembly machinery. Required for maturation of extramitochondrial Fe/S proteins. May bind and transfer a labile 4Fe-4S cluster to target apoproteins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NUBP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat