Anti-IKBIP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-7600R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:I Kappa B Kinase Interacting Protein
- Antigen symbol:IKBIP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:121457
- Antigen synonyms:IKIP|IKK Interacting Protein|IKBKB-Interacting Protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:270-305/350
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Target of p53/TP53 with pro-apoptotic function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IKBIP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human