Order Entry
Czech Republic
ContactUsLinkComponent
Anti-POLD2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-POLD2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-7416R-A488
Supplier:  Bioss
Anti-POLD2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-7416R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Polymerase (DNA Directed), Delta 2
  • Antigen symbol:
    POLD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    5425
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

DNA replication, recombination and repair, all of which are necessary for genomic stability, require the presence of exonucleases (1). In DNA replication, these enzymes are involved in the processing of Okazaki fragments, whereas in DNA repair, they function to excise damaged DNA fragments and correct recombinational mismatches (2). These exonucleases include the family of DNA polymerases (3). DNA pol α, β, ∂, and e are involved in DNA replication and repair (4). DNA pol ∂ and DNA pol e are multisubunit enzymes, with DNA pol ∂ consisting of two subunits p125, which interacts with the sliding DNA clamp protein PCNA, and p50 (5). The nuclear-encoded DNA pol © is the only DNA polymerase required for the replication of the mitochondrial DNA (6). DNA pol Ω is ubiquitously expressed in various tissues and mediates the cellular mechanism of damage-induced mutagenesis (7). DNA pol œ is a DNA polymerase-helicase that binds ATP and is involved in the repair of interstrand crosslinks (8).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POLD2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat