Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CDK19 Rabbit Polyclonal Antibody (Cy3®)
Anti-CDK19 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-7070R-CY3
Supplier:  Bioss
Anti-CDK19 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-7070R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent Kinase 19
  • Antigen symbol:
    CDK19
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    23097
  • Antigen synonyms:
    CDC2L6|bA346C16.3|CDK11
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a protein that is one of the components of the Mediator coactivator complex. The Mediator complex is a multiprotein complex required for transcriptional activation by DNA binding transcription factors of genes transcribed by RNA polymerase II. The protein encoded by this gene is similar to cyclin-dependent kinase 8 which can also be a component of the Mediator complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDK19
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: