Specifications
- Antibody type:Primary
- Antigen name:APAF1-interacting protein
- Antigen symbol:APIP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51074
- Antigen synonyms:MMRP19|CGI-29|CGI29|hAPIP|APIP2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The mammalian homologues of the key cell death gene CED 4 in C. elegans has been identified recently from human and mouse and designated Apaf1 (for apoptosis protease activating factor 1). Apaf1 binds to cytochrome c (Apaf2) and caspase 9 (Apaf3), which leads to caspase 9 activation. Activated caspase 9 in turn cleaves and activates caspase 3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. A new Apaf1 Interacting Protein (APIP) also known as CG129 and MMRP19, has been identified as a negative regulator of ischemic injury. APIP competes with Caspase 9 binding site of Apaf1. APIP is predicted to code for a 204 amino acid. An isoform of APIP, APIP2 encodes a 242 amino acid protein, which is an alternative splicing variant differing in its N terminus from APIP. APIP transcript is ubiquitously expressed in most adult tissue with high expression in skeletal muscle, heart, and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APIP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat