Anti-S100PBP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BOSSBS-6577R-HRP
: Bioss
- Antibody type:Primary
- Antigen name:S100P Binding Protein
- Antigen symbol:S100PBP
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Gene ID:64766
- Antigen synonyms:S100PBPR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:375-408/408
- Purification:Purified by Protein A
- Pk:100 µl
S100PBP was originally cloned from a pancreatic epithelioid carcinoma library and encodes a predicted 408 amino acid protein. RT-PCR detected S100PBP expression in brain, breast, spleen, and lung, but not in pancreas and liver. GFP-tagged S100PBP localized to nuclei of transfected HeLa cells.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: S100P Binding Protein
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat