Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ENTPD5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-ENTPD5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6526R-A488
Supplier:  Bioss
Anti-ENTPD5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6526R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ectonucleoside triphosphate diphosphohydrolase 5
  • Antigen symbol:
    ENTPD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    957
  • Antigen synonyms:
    PCPH|CD39L4|NTPDase-5
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    331-380/428
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Uridine diphosphatase (UDPase) that promotes protein N-glycosylation and ATP level regulation. UDP hydrolysis promotes protein N-glycosylation and folding in the endoplasmic reticulum, as well as elevated ATP consumption in the cytosol via an ATP hydrolysis cycle. Together with CMPK1 and AK1, constitutes an ATP hydrolysis cycle that converts ATP to AMP and results in a compensatory increase in aerobic glycolysis. The nucleotide hydrolyzing preference is GDP >IDP >UDP, but not any other nucleoside di-, mono- or triphosphates, nor thiamine pyrophosphate. Plays a key role in the AKT1-PTEN signaling pathway by promoting glycolysis in proliferating cells in response to phosphoinositide 3-kinase (PI3K) signaling.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENTPD5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat